Learn More
Clontech Labs 3P Wash and Stop Solution for ELISA without Sulfuric Acid | 5 x 96 Rxns
SDP
Supplier:  Clontech Labs 3P MK021
Encompass
This product contains 10X PBS and Tween20 for preparing the wash solution and Stop Solution for stopping the peroxidase reaction, which are used together with TaKaRa Bio' s ELISA Kits. Although the Stop Solution is not sulfuric acid (H2SO4) -based, it can be used like 1N H2SO4. Use the prepared wash solution and Stop Solution according to the user manual for each ELISA Kit.
Catalog No. NC1494269
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.