Learn More
Description
Includes
Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations
- High sensitivity
- High throughput, high content analysis
- Minimal volume of biological material required
- Suitable for frozen/thawed biological samples
- Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis
Specifications
Specifications
| Antigen | Granzyme B |
| Cross Reactivity | Human |
| Includes | Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates |
| Quantity | 10 x 96 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Cloning, proliferation assays, flow cytometry or other methods of analysis |
| Product Type | Human Granzyme B ELISPOT set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.