Learn More
Electron Microscopy Sciences ELITE Pad, 25/PK
SDP
Supplier:  Electron Microscopy Sciences 6333803
Encompass_Preferred
FAN Pad-GL ELITE is is a dry formalin neutralizer pad to absorb and neutralize excess formalin. Each pad is treated with a liquid formalin neutralizer, processed through a gentle drying process that maintains the same neutralizing effectiveness as a moist pad. The surface is cut-resistant and durable, providing an excellent grossing platform. The medium blue color cutting surface is a key visual benefit providing good contrast while grossing tissue, even when working with the smallest biopsies. Size: 15" x 20".
Catalog No. 50-209-0844
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.