Learn More
Fisherbrand™ Elite™ Multichannel Pipetters
SCI_ED
Catalog No. S98639A Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
5 to 50 μL
Number of Channels:
8
Catalog No. Volume (Metric) Number of Channels
S98639A 5 to 50 μL 8
1 options
This item is not returnable. View return policy
Catalog No. S98639A
This item is not returnable. View return policy
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort.
  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Number of Channels 8
Product Line Elite
Type Pipette
Volume (Metric) 5 to 50 μL
Warranty 3 years