Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics MGB Alert™™ Evaluation Sets

Catalog No. 22801050
Change view
Click to view available options
BK Virus 2.0 Primers (for evaluation)
BK Virus 2.0 Probes (for evaluation)
CMV 3.0 Primers (for evaluation)
EBV Primers (for evaluation)
EBV Probes (for evaluation)
HHV-6 Primers (for evaluation)
HHV-6 Probes (for evaluation)
HSV 1 and 2 Typer Primers (for evaluation)
HSV 1 and 2 Typer Probes (for evaluation)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-801-050 BK Virus 2.0 Primers (for evaluation)
22-801-052 CMV 3.0 Primers (for evaluation)
22-801-053 EBV Primers (for evaluation)
22-801-054 HHV-6 Primers (for evaluation)
22-801-055 HSV 1 and 2 Typer Primers (for evaluation)
22-801-056 BK Virus 2.0 Probes (for evaluation)
22-801-059 EBV Probes (for evaluation)
22-801-060 HHV-6 Probes (for evaluation)
22-801-061 HSV 1 and 2 Typer Probes (for evaluation)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-801-050 Supplier Nanogen, Inc Supplier No. 800411
This item is not returnable. View return policy

Primers, probes, and control sets are available in 48-reaction quantities for evaluation purposes

Specifications

Content And Storage 300μL Chemically synthesized DNA oligonucleotide primers
For Use With (Application) DNA Sequencing
No. of Reactions 48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.