Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics MGB Alert™ Primers

Catalog No. 22801070
Change view
Click to view available options
CMV 3.0 Primers
VZV Primers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-801-070 VZV Primers
22-801-067 CMV 3.0 Primers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-801-070 Supplier Nanogen, Inc Supplier No. 800418

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide molecular labs with a more advanced alternative to develop real-time PCR-based assays

Proprietary MGB™ and Superbase™ technologies allow post-PCR dissociated melt curve analysis, providing molecular labs with more powerful tools for detecting nucleic acid sequences of specific organisms associated with disease. MGB Alert real-time PCR reagents are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into existing workflow. Laboratories may use MGB Alert reagents to develop real-time PCR assays.

  • Compatible with most real-time PCR instruments
  • Optimized for universal cycling conditions
  • Easy to implement into existing workflow
  • Improved analysis from post-PCR melt curve capabilities

MGB Alert BK Virus Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the VP1 region of BK Virus
MGB Alert CMV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the IE1 region of CMV
MGB Alert CMV Q Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the IE1 region of CMV
MGB Alert EBV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the p140 region of the BNRF1 gene of EBV
MGB Alert Enterovirus Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the 5' UTR region of Enterovirus
MGB Alert HHV-6 Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the U67 region of HHV-6
MGB Alert hMPV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the Polymerase (L) gene of hMPV
MGB Alert HSV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the glycoprotein D region of HSV
MGB Alert HSV 1 and 2 Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the glycoprotein D region of HSV 1 and 2
MGB Alert MS2 Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the MS2 bacteriophage

Specifications

Content And Storage 30μL Chemically synthesized DNA oligonucleotide primers
For Use With (Application) DNA Sequencing
No. of Reactions 48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.