Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics MGB Alert™ Real-Time PCR Probes

Catalog No. 22801071
Change view
Click to view available options
CMV 3.0 Probes
CMV Probes
VZV Probes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-801-071 VZV Probes
22-801-068 CMV Probes
22-801-058 CMV 3.0 Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-801-071 Supplier Nanogen, Inc Supplier No. 800419

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide molecular labs with a more advanced alternative to develop real-time PCR-based assays

Proprietary MGB™ and Superbase™ technologies allow post-PCR dissociated melt curve analysis, providing molecular labs with more powerful tools for detecting nucleic acid sequences of specific organisms associated with disease. MGB Alert real-time PCR reagents are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into existing workflow. Laboratories may use MGB Alert reagents to develop real-time PCR assays.

  • Compatible with most real-time PCR instruments
  • Optimized for universal cycling conditions
  • Easy to implement into existing workflow
  • Improved analysis from post-PCR melt curve capabilities

MGB Alert BK Virus Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from the VP1 region of BK Virus
MGB Alert CMV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from CMV
MGB Alert EBV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from EBV
MGB Alert Enterovirus Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from Enterovirus
MGB Alert HHV-6 Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from HHV-6
MGB Alert hMPV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from the Polymerase (L) gene of hMPV
MGB Alert HSV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from HSV
MGB Alert HSV 1 and 2 Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from HSV 1 and 2
MGB Alert MS2 Probes and MS2-E Probes consist of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from the MS2 bacteriophage

Specifications

Content And Storage 30μL Chemically synthesized DNA oligonucleotide probe
For Use With (Application) DNA Sequencing
No. of Reactions 48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.