Learn More
Invitrogen™ ELK1 Recombinant Rabbit Monoclonal Antibody (K01_1Q74)

Description
ELK1 Recombinant Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ELK1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1Q74 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | ELK1 |
| Gene Accession No. | A4GTP4, P19419, P41969 |
| Gene Alias | ELK 1; ELK1; Elk-1; ELK1, ETS transcription factor; ELK1, member of ETS oncogene family; ETS domain-containing protein Elk-1; ETS transcription factor ELK1; ETS-like gene 1; Oncogene Elk1; tyrosine kinase (ELK1) oncogene |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.