Learn More
Sigma Organic Chemistry Ellagic acid hydrate | 10G
Supplier: Sigma Organic Chemistry 37274910G

Ellagic acid hydrate, 10G
About This Item:
Linear Formula: C14H6O8 -+ xH2O
UNSPSC Code: 12162002
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.