Learn More
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, N2a, PC-12, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ELMO1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ELMO1 |
| Gene Accession No. | Q8BPU7, Q92556 |
| Gene Alias | 6330578D22Rik; C230095H21Rik; CED12; CED-12; ced-12 homolog 1; ELMO1; ELMO-1; ELMO1s; engulfment and cell motility 1; engulfment and cell motility 1 (ced-12 homolog, C. elegans); engulfment and cell motility 1 isoform 2; engulfment and cell motility 1 splice 1; engulfment and cell motility 1, ced-12 homolog; engulfment and cell motility protein 1; im:7151147; KIAA0281; MGC126406; OTTHUMP00000207056; OTTHUMP00000207060; Protein ced-12 homolog; wu:fq24e08; zgc:85612 |
| Gene Symbols | ELMO1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
