Learn More
Aviva Systems Biology Corp ELMOD2 Antibody - N-terminal region (ARP55604_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55604_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELMOD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ELMOD2. Alternate names: 9830169G11Rik, MGC10084ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61552