Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELOVL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592039 Supplier Invitrogen™ Supplier No. PA592039
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGLLDSEPGS VLNVVSTALN DTVEFYRWTW SIADKRVENW PLMQSPWPTL SISTLYLLFV WLGPKWMKDR EPFQMRLVLI IYNFGMVLLN LFIFRELFMG; Positive Samples: U-87MG, HT-29, OVCAR3, A-431, HeLa; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

This gene encodes a membrane-bound protein which is a member of the ELO family, proteins which participate in the biosynthesis of fatty acids. Consistent with the expression of the encoded protein in photoreceptor cells of the retina, mutations and small deletions in this gene are associated with Stargardt-like macular dystrophy (STGD3) and autosomal dominant Stargardt-like macular dystrophy (ADMD), also referred to as autosomal dominant atrophic macular degeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOVL4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ELOVL4
Gene Accession No. Q9EQC4, Q9GZR5
Gene Alias 3-keto acyl-CoA synthase Elovl4; ADMD; cancer/testis antigen 118; CT118; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4; elongation of very long chain fatty acids protein 4; elongation of very long chain fatty acids-like 4; ELOVL FA elongase 4; ELOVL fatty acid elongase 4; Elovl4; FLJ17667; FLJ92876; ISQMR; SCA34; STGD2; STGD3; Very long chain 3-ketoacyl-CoA synthase 4; very long chain 3-oxoacyl-CoA synthase 4; very-long-chain 3-oxoacyl-CoA synthase 4
Gene Symbols ELOVL4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315851, 6785, 83603
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.