Learn More
3M™ Scott™ ELSA™ Emergency Escape Breathing Device - N (EEBD-N)
Description
Enables workers to escape from certain toxic environments, even in atmospheres immediately dangerous to life or health (IDLH).
- Features one-size-fits-all clear hood with patent-pending neck seal
- Pulls on with no additional steps to secure, making it easy to train and use properly
- Includes fully-enclosed, chemical-resistant PVC carry bag to keep it free of insects and debris
- Available with 5, 10, or 15 minute durations
- Available with full or drained cylinders
- NIOSH approved
- 10 and 15 minute models meet SOLAS Chapter II-2, Part D requirements.
- Fits into standard wall cases, has indefinite shelf life, and comes with a 3 year warranty.
Specifications
Specifications
| Certifications/Compliance | NIOSH, SOLAS Chapter II-2 Part D |
| Product Type | Escape Breathing Apparatus |
| Duration | 15 min. |
| Pressure | 30 lpm |
| Hood Type | Clear hood with patent-pending neck seal fits all wearers, and pulls on with no additional steps to secure, making it easy to train and use properly. |
| Includes | Respirator, Hood, Carry Bag, Full Cylinder |
| Model | 8007214 |