Learn More
3M™ Scott™ ELSA™ Emergency Escape Breathing Device - N (EEBD-N)
Catalog No. 19072402 Shop All 3M Products
Click to view available options
Certifications/Compliance:
NIOSH
NIOSH, SOLAS Chapter II-2 Part D
Includes:
Respirator, Hood, Carry Bag, Drained Cylinder
Respirator, Hood, Carry Bag, Full Cylinder
Duration:
10 min.
15 min.
5 min.
Catalog No. 19-072-402
Supplier: 3M™ Scott™ 8007214D
ELSA™ EEBD-N respirator emergency escape breathing device is designed for simple use, training, and maintenance. Carry bag offers improved functionality while the redesigned hood fits wearers comfortably.
Enables workers to escape from certain toxic environments, even in atmospheres immediately dangerous to life or health (IDLH).
  • Features one-size-fits-all clear hood with patent-pending neck seal
  • Pulls on with no additional steps to secure, making it easy to train and use properly
  • Includes fully-enclosed, chemical-resistant PVC carry bag to keep it free of insects and debris
  • Available with 5, 10, or 15 minute durations
  • Available with full or drained cylinders
  • NIOSH approved
  • 10 and 15 minute models meet SOLAS Chapter II-2, Part D requirements.
  • Fits into standard wall cases, has indefinite shelf life, and comes with a 3 year warranty. 
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH, SOLAS Chapter II-2 Part D
Product Type Escape Breathing Apparatus
Duration 15 min.
Pressure 30 lpm
Hood Type Clear hood with patent-pending neck seal fits all wearers, and pulls on with no additional steps to secure, making it easy to train and use properly.
Includes Respirator, Hood, Carry Bag, Drained Cylinder
Model 8007214-D
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.