Learn More
Invitrogen™ ELSPBP1 Polyclonal Antibody

Description
Immunogen sequence: MTRWSSYLLG WTTFLLYSYE SSGGMHEECV FPFTYKGSVY FTCTHIHSLS PWCATRAVYN GQWKYCQSED YPRCIFPFIY RGKAYNSCIS QGSFLGSLWC SVTSVFDEKQ QWKFCETNEY GGNSLRKPCI FPSIYRNNVV SDCMEDESNK LWCPTTENMD KDGKWSFCAD TRISALVPGF PCHFPFNYKN KNYFNCTNEG SKENLVWCAT SYNYDQDHTW VYC; Positive Samples: HepG2, 293T.
Specifications
Specifications
| Antigen | ELSPBP1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ELSPBP1 |
| Gene Accession No. | Q96BH3 |
| Gene Alias | E12; EDDM12; EL149; ELSPBP1; epididymal protein 12; epididymal secretory protein 12; epididymal sperm binding protein 1; epididymal sperm-binding protein 1; epididymis luminal secretory protein 149; ESPB1; hE12 |
| Gene Symbols | ELSPBP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.