Learn More
Apexbio Technology LLC SB408124 288150-92-5 5mg
SDP
Supplier:  Apexbio Technology LLC B21635
Encompass_Preferred
SB408124 (CAS 288150-92-5) is a non-peptide antagonist of the orexin-1 receptor (OX1) It displays receptor selectivity exhibiting approximately 50-fold greater affinity for OX1 compared to OX2 receptors in humans with reported Ki values of 27 nM (membrane preparations) and 57 nM (whole-cell assays) Experimental studies using primary cultured rat astrocytes indicate that SB408124 pretreatment can attenuate orexin-A-mediated stimulation of cAMP production In vivo rat models demonstrate that co-administration with orexin-A prevents reductions in vasopressin elevations induced by histamine or hyperosmotic challenge SB408124 serves as a tool compound in research exploring the physiological roles of OX1 receptors in neuroendocrine and cellular signaling pathways
Catalog No. 50-000-62428
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.