Learn More
Apexbio Technology LLC Eltrombopag Olamine 496775-62-3 200mg
SDP
Supplier:  Apexbio Technology LLC A3387200
Encompass_Preferred
Eltrombopag Olamine is an orally bioavailable thrombopoietin receptor (c-mpl) agonist that directly stimulates platelet production through activation of the JAK2/STAT signaling pathway In cellular assays using murine BAF3 cells expressing human thrombopoietin receptor (hTpoR) eltrombopag induced STAT-dependent signaling responses with an EC50 value of approximately 0 27 M This compound promotes differentiation of human CD34 progenitor cells into megakaryocytes capable of platelet production Eltrombopag is commonly applied in hematological research particularly in studies involving thrombocytopenia and thrombopoiesis-related mechanisms
Catalog No. 50-000-62832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.