Learn More
Thermo Scientific Filter Frit (10μm Porosity)
Supplier:  Thermo Scientific 6268.0111
Encompass
Filter Frit (10μm Porosity), titanium, for use with Ultimate 3000 SD, RS Pumps
Catalog No. 50-134-8075
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.