Learn More
Eluent Solution, for Ion Chromatography, 3 mM NaHCO3 - 2.4 mM Na2CO3, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Eluent Solution |
| Color | Colorless |
| Name Note | 3mM NaHCO |
| CAS | 7732-18-5, 497-19-8, 144-55-8 |
| CAS Min % | 98.0, 0.02, 0.02 |
| CAS Max % | 100.0, 0.03, 0.03 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Quantity | 4 L |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.