Learn More
Glass Expansion Eluo Adaptor for Inline Filter
SDP
Supplier:  Glass Expansion 708031160
Encompass_Preferred
Eluo Adaptor for In-Line Filter. This adaptor is designed to assist in backflushing Glass Expansion’s Guardian In-Line Filter (NC0897716). It is used in conjunction with the barrel (NC1906608) and plunger (NC1906609) which are already supplied with either the Eluo Nebulizer Cleaner for Glass Concentric Nebulizers (NC9462559) or the Eluo Nebulizer Cleaner for OpalMist and DuraMist Nebulizers (501907353). If you already have an Eluo Nebulizer Cleaning Tool, you can simply remove the nebulizer adaptor and replace it with the in-line filter adaptor to backflush the Guardian In-Line Filter.Fluka RBS-25 Concentrate 1L (NC9000117) is an excellent cleaning solution for nebulizers and other sample introduction components.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC0801128