Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience Elution Buffers for G8 Automated Glycohemoglobin Analyzer
For use with G8 Automated Glycohemoglobin Analyzer.
Specifications
For Use With (Equipment) | G8 automated HPLC glycohemoglobin analyzer |
---|---|
Quantity | 800 mL |
Content And Storage | 4°C to 30°C |
Description
- For use with G8 Automated Glycohemoglobin Analyzer
- Quantity: 800mL
Specifications
G8 automated HPLC glycohemoglobin analyzer | |
800 mL | |
4°C to 30°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title