Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Supplements

Catalog No. SR0258E
Change view
Click to view available options
Yield:
1 vial / 500 mL agar
Form:
Liquid
Pellets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield Form
SR0258E 1 vial / 500 mL agar Liquid
SR0257E 1 vial / 500 mL agar Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SR0258E Supplier Thermo Scientific™ Supplier No. SR0258E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enable the performance of Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) with these selective and differential supplements.

Enable the performance of Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Base with the Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Supplements.

When used to supplement the Brilliance™ Listeria Agar (ISO) Supplements, the Selective Supplement enables the inhibition of competing microflora whilst the Differential Supplement aids the differentiation of Listeria monocytogenes from other Listeria species by facilitating the development of an opaque white halo around the colonies.

Enable the performance of the Thermo Scientific™ Oxoid™ Brilliance™ Listeria (ISO) Agar with the Brilliance™ Listeria (ISO) Agar Supplements.

  • Available in convenient sizes suitable for supplementing 5 L of Brilliance Listeria (ISO) Agar (E pack size) or compatible with our 500 mL bottled media options for preapring Pour plates (B pack size).
  • Each box of supplement contains 10 vials providing a convenient way to make as much or as little Brilliance Listeria (ISO) Agar as you need.
  • Brilliance Listeria (ISO) Agar is intended to be used as part of the Thermo Scientific™ Oxoid™ Listeria Precis™ Methods or  as part of the ISO 11290-1:2017 and ISO 11290-2:2017 reference methods for the detection and enumeration of Listeria species and L. monocytogenes.
  • The Listeria Precis Methods are validated for a broad range of samples according to ISO 16140-2 following NF VALIDATION rules when compared to the ISO 11290-1:2017 and ISO 11290-2:2017 Microbiology of the food chain – Horizontal methods for the detection and enumeration of Listeria monocytogenes and of Listeria spp.
TRUSTED_SUSTAINABILITY

Specifications

Description Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO)Differential Supplement (Cat. No. SR0258E)
Form Liquid
Format Box of 10 vials
Product Type Media Supplement
Formulation L-α-phosphatidylinositol solution
Yield 1 vial / 500 mL agar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.