Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ E. coli DNA Ligase

Catalog No. FEREL00361 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREL00361 400 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREL00361 Supplier Thermo Scientific™ Supplier No. EL00361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ E. coli DNA Ligase is an NAD-dependent DNA ligase. It catalizes formation of phosphodiester bonds between 5'-phospate and 3'-hydroxyl termini in double-stranded DNA.

Thermo Scientific™ E. coli DNA Ligase is an NAD-dependent DNA ligase. It catalizes formation of phosphodiester bonds between 5'-phospate and 3'-hydroxyl termini in double-stranded DNA. The enzyme is not active on blunt-ended DNA.

Features

  • Activity in wide temperature range (4°C to 37°C)
  • High specificity to cohesive DNA ends

Applications

  • Nick ligation in double-stranded DNA
  • cDNA synthesis
  • Cloning

TRUSTED_SUSTAINABILITY

Specifications

Concentration 8 U/μL
For Use With (Application) Nick ligation in double-stranded DNA, cDNA synthesis, Cloning, DNA ligation
Content And Storage 8 U/μL E.Coli DNA Ligase, 50 μL

Store at -20°C.
Enzyme DNA Ligase
Sensitive to Heat Inactivation Yes
Quantity 400 U
Product Type NAD-dependent DNA Ligase
Research Category Traditional Cloning
Inactivation Temperature 65°C
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.