Learn More
Apexbio Technology LLC Embelin, 10mg. Cas: 550-24-3 MFCD: MFCD00016369. XIAP inhibitor,cell-permeable
SDP
Supplier:  Apexbio Technology LLC A823510
Encompass_Preferred
Embelin, a natural benzoquinone from plants of the genus Embelia, is an inhibitor of X-linked inhibitor of apoptosis protein (XIAP) with IC50 of 4.1 M.XIAP is a member of the inhibitor of apoptosis family of proteins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5088