Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Embigin Monoclonal Antibody (G7.43.1), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5010899
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-108-99 100 μg
50-108-98 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-108-99 Supplier Invitrogen™ Supplier No. 12583982
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This G7.43.1 monoclonal antibody reacts with mouse embigin (gp70), a single transmembrane glycoprotein that is a member of the Ig superfamily similar to basigin and EMMPRIN. It contains two Ig domains thought to be involved in cellular adhesion and tissue remodeling. Expression of embigin is high before day 10 in fetal development. In the adult it is expressed on myeloid cells and T cells but not B cells. During hematopoeisis, embigin is expressed on HSCs and CLPs (common lymphoid progenitors) but absent on pro- and pre-B cells. In recent studies Pax 5 has been shown to repress embigin expression. Applications Reported: This G7.43.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G7.43.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Embigin is a 327 amino acid regulatory protein belonging to the immunoglobulin superfamily class of CAMs, and has two Ig-like (immunoglobulin-like) V- type domains. In situ hybridization analysis shows that Embigin has specific carbohydrate epitopes and plays an important role in early postimplantation embryogenesis. A highly glycosylated transmembrane glycoprotein, Embigin acts as a regulator of cell/ECM interactions during development and homeostasis of normal adult tissues. Transfection studies suggest its role in promotion of integrin-mediated cell-substratum adhesion. Widely expressed in most of the tissues and embryonal carcinoma cells, prostate and mammary cancer cell lines, it may also help in the classification of human carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Embigin
Applications Flow Cytometry
Classification Monoclonal
Clone G7.43.1
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene EMB
Gene Accession No. P21995
Gene Alias AL022799; EMB; Embigin; embigin homolog; glycoprotein 70; Gp70; Teratocarcinoma glycoprotein Gp-70
Gene Symbols EMB
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13723
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.