Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EMD 281014 hydrochloride

Catalog No. 447050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-705-0 50 mg
44-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-705-0 Supplier Tocris Bioscience™ Supplier No. 4470/50
This item is not returnable. View return policy

Selective 5-HT2A antagonist

Selective 5-HT2A antagonist; displays high affinity for human 5-HT2A over 5-HT2C receptors (Ki values are 0.87 and 557 nM respectively). Exhibits low off-target activity (Ki values are >1000 and 1268 nM for human D2 receptors and IKr channels, respectively).

Chemical Identifiers

CAS 443144-27-2
Molecular Formula C22H22ClFN4O

Specifications

Quantity 50 mg
Formula Weight 412.89
Purity >98%
Molecular Weight (g/mol) 412.893
Chemical Name or Material EMD 281014 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.