Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EMD 386088 hydrochloride
GREENER_CHOICESDP

Catalog No. 238250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-825-0 50 mg
23-821-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-825-0 Supplier Tocris Bioscience™ Supplier No. 2382/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent 5-HT6 agonist

EMD 386088 hydrochloride is a potent 5-HT6 receptor agonist (EC50 = 1.0 nM) that displays selectivity over other 5-HT receptors (IC50 values are 7.4, 110, 180, 240, 450, 620, 660 and 3000 nM for 5-HT6, 5-HT1D, 5-HT1B, 5-HT2A, 5-HT2C, 5-HT4, 5-HT1A and 5-HT7 receptors respectively). Shows moderate affinity at 5-HT3 receptors (IC50 = 34 nM).

Specifications

Chemical Name or Material 5-Chloro-2-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole hydrochloride
CAS 1171123-46-8
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C14H15N2Cl.HCl
Formula Weight Observed MW: 283.2

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.