Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α1,2-Fucosidase, Xanthomonas sp., Protein

Catalog No. 3448225MIU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-482-25MIU 5 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-482-25MIU Supplier MilliporeSigma™ Supplier No. 3448225MIU
Add to Cart
Add to Cart
Native α1,2-fucosidase from Xanthomon as species. Catalyzes the hydrolysis of all non-reducing terminal fucose that are α1,2-linked to the galactose of terminal Gal-GlcNAc disaccharide structures. The enzyme will not cleave fucose linked to the GlcNAc residue.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 25mM NaCl, 20mM Tris-HCl, pH 7.5.
Quantity 5 mIU
Source Xanthomonas sp.
Immunogen α1,2-Fucosidase
Regulatory Status RUO
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.