Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ β1-3,6-Galactosidase, Recombinant Protein, E. coli

Catalog No. 3458042U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-580-42U 2 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-580-42U Supplier MilliporeSigma™ Supplier No. 3458042U
Add to Cart
Add to Cart
Recombinant, β1-3,6-Galactosidase expressed in E. coli. Catalyzes the hydrolysis of non-reducing terminal β1,3- and β1,6-linked galactose from complex carbohydrates and glycoproteins. Not known to cleave substituted or branched galactose. Inhibited by fucose attached to penultimate sugar.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 25mM NaCl, 20mM Tris-HCl, pH 7.5.
Quantity 2 U
Source E. coli
Immunogen β1-3,6-Galactosidase
Regulatory Status RUO
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.