Learn More
MilliporeSigma™ β1,4-Galactosidase, Streptococcus pneumoniae, Recombinant Protein, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 25mM NaCl, 20mM Tris-HCl, pH 7.5. Sterile solution. |
| Quantity | 50 mIU |
| Source | E. coli |
| Immunogen | β1,4-Galactosidase |
| Regulatory Status | RUO |
| Species | S. pneumoniae |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.