Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α2-3,6-Neuraminidase, Clostridium perfringens, Recombinant Protein, E. coli

Catalog No. 480708500MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-070-8500MI 500 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-070-8500MI Supplier MilliporeSigma™ Supplier No. 480708500MIU
Add to Cart
Add to Cart
Recombinant, Clostridium perfringens α2-3,6-Neuraminidase expressed in E. coli. Catalyzes the hydrolysis of non-reducing terminal α2,3 and α2,6 unbranched sialic acid residues from complex carbohydrates and glycoproteins. This enzyme does not exhibit activity on α2,8 or branched sialic acids.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 25mM NaCl, 20mM Tris-HCl, pH 7.5.
Quantity 500 mIU
Source E. coli
Immunogen α2-3,6-Neuraminidase
Regulatory Status RUO
Species C. perfringens
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.