Learn More
MilliporeSigma™ β2-Microglobulin, Human, Recombinant Protein, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Preparation Method | Aqueous buffers |
| Purification Method | Single band by SDS-PAGE |
| Quantity | 1 mg |
| Source | E. coli |
| Immunogen | β2-Microglobulin |
| Storage Requirements | Following initial thaw, aliquot and freeze (−20°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.