Learn More
Description
Specifications
Specifications
| Formulation | In PBS, 150mM NaCl, 20mM phosphate buffer, pH 7.3. |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Immunogen | β2-Microglobulin |
| Storage Requirements | Following initial thaw, product can be stored at 4°C for up to 6 months. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
| Purity or Quality Grade | ≥98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.