Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ 384-Well PI3 Kinase Homogenous Cell-Based Assay Kits
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Description | 384-Well PI3 Kinase Homogenous Cell-Based Assay Kit |
| Detection Method | TR-Fret HTRF |
| For Use With (Application) | For assaying PIP3 levels by detecting the displacement of biotin-PIP3 from an energy transfer complex |
| Includes | 1M DTT,Black 384-well plate,Detection Buffer,DM B,DM A,Stop B,PIP3 Standard,Lipid Recovery Buffer |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?