Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hydrosol Stainless Steel Filter Holder and Accessories
GSA_VA

Catalog No. XX2004720 Shop All MilliporeSigma Products
Change view
Click to view available options
Volume (Metric):
650 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
XX2004720 650 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX2004720 Supplier MilliporeSigma™ Supplier No. XX2004720
Add to Cart
Add to Cart

Hydrosol Stainless Steel Filter Holder and Accessories for particulate or biological contamination analysis via vacuum filtration.

XX2004720 Hydrosol Filter Holder, 47 mm, stainless steel:

  • The Hydrosol filter holder is designed for the filtration of flammable liquids.
  • Unlike other filter holders, the Hydrosol filter holder includes a grounding unit to prevent sparking from static discharge, which can be a safety hazard.
  • Includes grounding set for use with flammable liquids, stainless-steel funnel, base, and filter support screen, anodized aluminum locking ring, nylon lockwheels, PTFE resin gaskets, silicone stopper.

Applications: Vacuum Filtration of Flammable Liquids

Replacement Parts:

  • XX2004704 Funnel and Locking Ring Assembly, stainless steel
  • XX2004708 Support Screen, 47 mm, stainless steel
  • XX2004702 Base with Screen, gasket and support
  • XX2004703 Gasket, PTFE
  • XX2004718 No. 8 Stopper, 9.5 mm (3/8 in.) hole, silicone
  • XX20047RK Replacement Parts Kit for Hydrosol Filter Holder Accessories
  • XX1004705 Vacuum Filtering Flask, 1 L
  • XX1004744 Vacuum Filtering Flask, 4 L
  • XX7100004 Tubing 3/16 in. I.D. x 140 cm, silicone
  • XX6200006P Filter Forceps, blunt end, stainless steel
  • XX2604735 3-place Manifold, 47 mm, PVC
  • XX2504735 3-place Manifold, 47 mm, stainless steel
  • XX2504700 6-place Manifold, 47 mm, stainless steel
  • WP6211560 High Output Pump, 115 V/60 Hz
  • WP6222050 High Output Pump, 220 V/50 Hz
  • WP6210060 High Output Pump, 100 V/50-60 Hz

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Disposable Reusable
Material Stainless Steel
Sterility Non-sterile
Type Filter Holder
Volume (Metric) 650 mL
Height (Metric) 22.9 cm
Diameter (English) Inside 4.5 in.
Diameter (Metric) Filter 47 mm
Diameter (Metric) Inside 11.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.