Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ 5-Lipoxygenase, Human, Recombinant Protein, S. fruigiperda

Catalog No. 437996500U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-799-6500U 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-799-6500U Supplier MilliporeSigma™ Supplier No. 437996500U
Add to Cart
Add to Cart
Recombinant, human 5-Lipoxygenase expressed in S. fruigiperda insect cells using a baculovirus overexpression system. 5-LO catalyzes the formation of 5(S)-HpETE from arachidonic acid and its subsequent conversion to leukotriene A4. It is localized in the cytosol of resting human and rat peripheral blood neutrophils, whereas it is found in the nucleus in rat basophilic leukemia cells and human alveolar macrophages. Upon stimulation of the cell, 5-LO translocates to the nuclear membrane where it is found in association with 5-LO activating protein (FLAP) and with the 85kDa cPLA2.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM Tris containing 5mM EGTA, pH 8.0.
Quantity 500 U
Source S. fruigiperda
Immunogen 5-Lipoxygenase
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.