Learn More
MilliporeSigma™ Inline filter cartrige holder.
Catalog No. M1511930001 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
M1511930001 1/Pk.
1 options
Catalog No. M1511930001 Supplier MilliporeSigma™ Supplier No. 1.51193.0001
Inline filter cartrige holder.
TRUSTED_SUSTAINABILITY
Specifications
Type In-Line Filter Holder
Packaging Quantity 1/Pk.
For Use With LiChrospher ADS
Particle Size 25 μm