Learn More
MilliporeSigma™ Gaskets with Peek Filter or LiChrospher™ ADS
Catalog No. M1511920001 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
M1511920001 5/Pk.
1 options
Catalog No. M1511920001 Supplier MilliporeSigma™ Supplier No. 1.51192.0001
5 gaskets with PEEK filter for LichroCART™ ADS LiChroCART™
Inline- Filtercartridge contains 5 pieces gasket ring.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement In-Line Filter
Packaging Quantity 5/Pk.
For Use With LiChrospher ADS
Particle Size 25 μm