Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Androgen Receptor, HisTag™, Human, Recombinant Protein, S. frugiperda

Catalog No. 3461015000U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-610-15000U 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-610-15000U Supplier MilliporeSigma™ Supplier No. 3461015000U
Add to Cart
Add to Cart
Recombinant, human androgen receptor (AR) fused at the N-terminus to a Flag-Tag™ sequence and at the C-terminus to His·Tag™ sequence, expressed in S. frugiperda insect cells expressed using a baculovirus expression system, and purified by affinity chromatography. The activity of androgen is mediated by the binding of the intracellular androgen receptor which, when activated, translocates to the nucleus and modulates specific gene transcription by interaction with cis-acting hormone response elements in the target genes.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM KCl, 20mM Tris-HCl, 1mM DTT, 200μM EDTA, 20% glycerol, pH 8.0.
Quantity 5000 U
Source S. frugiperda
Immunogen Androgen Receptor
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.