Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Angiotensinogen, Human Plasma, Protein

Catalog No. 176870100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-687-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-687-0100UG Supplier MilliporeSigma™ Supplier No. 176870100UG
Add to Cart
Add to Cart
Native angiotensinogen from human plasma. An α2-globulin present in plasma at about 7mg/100mL. Can be cleaved by renin to generate angiotensin I.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 50mM sodium phosphate, 150mM NaCl, pH 7.0.
Preparation Method H2O. Please refer to vial label for lot-specific concentration prior to lyophilization.
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen Angiotensinogen
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Solid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.