Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Apolipoprotein A-II, Human Plasma, High-Density Lipoprotein, Protein

Catalog No. 178455500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-845-5500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-845-5500UG Supplier MilliporeSigma™ Supplier No. 178455500UG
Add to Cart
Add to Cart
Native apolipoprotein A-II from human plasma. Shown to bind to phospholipids during lipoprotein metabolism. Displaces lecithin-cholesterol acyltransferase bound to lipoprotein. Influences HDL functional states and contributes to arteriosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 10mM NH4HCO3, pH 7.4.
Purification Method SDS-PAGE
Quantity 500 μg
Immunogen Apolipoprotein A-II
Storage Requirements Following initial thaw, aliquot and freeze (−20°C). Store undiluted.
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.