Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Apolipoprotein C-III, Human Plasma, Very Low-Density Lipoprotein, Protein

Catalog No. 178461100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-846-1100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-846-1100UG Supplier MilliporeSigma™ Supplier No. 178461100UG
Add to Cart
Add to Cart
Native apolipoprotein C-III from human plasma. Major protein of VLDL and chylomicrons. Involved in the uptake of triglycerides by cells. May inhibit the activation of lipoprotein lipase by Apo C-II. Present in normal plasma at 80-150μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 10mM NH4HCO3, pH 7.4.
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen Apolipoprotein C-III
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.