Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Content And Storage | Room temperature in clean dry area away from any chemicals or direct light |
| Description | AXIS Axon Isolation Device, Plasma Bonded |
| For Use With (Application) | Immunocytochemistry, Cell Culture |
| Includes | 5 units of AX450, axon isolation device consisting of four wells, two channels, and set of microgrooves |
| Quantity | 5 Devices |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?