Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BAFF Human Recombinant Protein

Catalog No. GF136 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF136 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF136 Supplier MilliporeSigma™ Supplier No. GF136
Add to Cart
Add to Cart

Specifications

For Use With (Application) Cell Culture
Name Human Recombinant Protein, BAFF
Quantity 20 μg
Source E. coli
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade Greater than 98% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.