Learn More
MilliporeSigma™ BioPak™ Disposable Ultrafilter
GSA_VA
Catalog No. CDUFBI001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
CDUFBI001 1 Pk.
1 options
Catalog No. CDUFBI001 Supplier MilliporeSigma™ Supplier No. CDUFBI001
Add to Cart
Add to Cart
Nuclease-free, pyrogen-free water
  • Guaranteed nuclease-free and pyrogen-free performance for three months, regardless of volumes used
  • Disposable filter eliminates the need for regular, lengthy sanitization procesures and hazardous chemicals
  • Fits all lab-scale EMD Millipore Ultrapure water systems: Simplicity portable water systems, Synergy™ ultrapure water systems, Direct-Q™ water systems, and all generations of Milli-Q™ water systems
  • Preserves water quality as well as performance—no loss in system flowrate
  • Cost-effective upgrade for your existing EMD Millipore ultrapure water system
Applications:
Biological techniques requiring nuclease-free and/or pyrogen-free water
Replacement of molecular biology-grade and cell culture-grade bottled water with similar performance
Advanced life science research
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Filter Type Ultrafiltration
For Use With (Application) Production of pyrogen-free and RNase-free water
For Use With (Equipment) EMD Millipore™ Ultrapure™, Synergy™ Ultrapure™, Direct-Q™ and all generations of Milli-Q™ water systems
Quantity 1 Pk.
Type Ultrafiltration Cartridge