Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ C-Reactive Protein, Human, Recombinant Protein, E. coli

Catalog No. 2366081MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-660-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-660-81MG Supplier MilliporeSigma™ Supplier No. 2366081MG
Add to Cart
Add to Cart
Recombinant, human C-reactive protein expressed in E. coli. CRP is a major acute-phase plasma protein that is dramatically elevated in patients with acute-phase conditions. Composed of five identical noncovalently linked subunits. Exhibits Ca2+-dependent binding to a variety of substrates. Important in diagnosis of rheumatoid arthritis. Suitable for immunological studies. In purified states and in plasma the monomeric form (M.W. ∽21kDa) aggregates as a cyclic pentamer.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 140mM NaCl, 20mM Tris-HCl, 2mM CaCl2, 0.05% NaN3, pH 7.5.
Preparation Method Aqueous buffers
Quantity 1 mg
Source E. coli
Immunogen C-Reactive Protein
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.