Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ C-Reactive Protein, Human Serum, High Purity, Protein

Catalog No. 236603100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-660-3100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-660-3100UG Supplier MilliporeSigma™ Supplier No. 236603100UG
Add to Cart
Add to Cart
Native C-reactive protein highly purified from human serum. CRP is a major acute-phase plasma protein that is dramatically elevated in patients with acute-phase conditions. Composed of five identical noncovalently linked subunits. Exhibits Ca2+-dependent binding to a variety of substrates. Important in diagnosis of rheumatoid arthritis. Suitable for immunological studies. In purified states and in plasma the monomeric form (M.W. ∽21kDa) aggregates as a cyclic pentamer. Intended for use as a calibration standard for assays of CRP, for structural and functional studies, and as an antigen for production of antisera. Calibrated by electroimmunoassay against WHO reference standard or equivalent.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 140mM NaCl, 10mM Tris, 2mM CaCl2, 0.1% NaN3, pH 8.0.
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen C-Reactive Protein
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥99%, High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.