Learn More
MilliporeSigma™ Calbiochem™ 20S Proteasome Assay Kit, SDS-Activated
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Content And Storage | Upon arrival of the kit store the Enzyme Solution -80°C. The remainder of the kit should be stored at -20°C |
| Description | 20S Proteasome Assay Kit, SDS-Activated |
| Detection Method | Fluorometric |
| For Use With (Application) | Measures 20S proteasome activity |
| Includes | 20S Proteasome in Buffer, Reaction Buffer, SDS, Substrate Solution and user protocol |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.