Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Avidin, Egg White

Catalog No. 189725100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-972-5100MG 100 mg
18-972-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-972-5100MG Supplier MilliporeSigma™ Supplier No. 189725100MG
Add to Cart
Add to Cart

Glycoprotein containing four identical subunits

  • Form: White lyophilized solid
  • Molar Mass: 66,000
  • Biological Activity: ≥10 units/mg protein
  • Soluble in dH2O and dilute buffer
  • Storage: Store at -20°C
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.