Learn More
Description
- Form: Lyophilized orange solid
- Soluble in DMF, DMSO, or MeOH
- Applications: Enzyme Immunoassay, Immunohistochemistry
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | Biotin, Fluorescein Conjugate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.